Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AlphaB Crystallin/CRYAB, p Ser45 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB1205598 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1205598 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1205598 Supplier Novus Biologicals Supplier No. NB1205598

Rabbit Polyclonal Antibody

AlphaB Crystallin/CRYAB Polyclonal specifically detects AlphaB Crystallin/CRYAB in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.

Specifications

Antigen AlphaB Crystallin/CRYAB (p Ser45)
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/ml, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin 8 ug/ml, Immunofluorescence 5 ug/ml
Formulation PBS with 0.05% Sodium Azide
Gene Alias alpha B crystallin, alpha(B)-crystallin, alpha-crystallin B chain, CRYA2alpha crystallin B chain, crystallin, alpha B, CTPP2, Heat shock protein beta-5, heat-shock 20 kD like-protein, HSPB5, Renal carcinoma antigen NY-REN-27, Rosenthal fiber component
Gene Symbols CRYAB
Host Species Rabbit
Immunogen Synthetic phopshopeptide corresponding to residues L(44) S(p) P F Y L R P P S F(54) C of human alpha-B Crystallin.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience, Phospho Specific, Vision
Primary or Secondary Primary
Gene ID (Entrez) 1410
Test Specificity This does not detect the unphosphorylated form of the protein.
Reconstitution Reconstitute to desired concentration.
Target Species Human, Mouse, Rat, Bovine
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.