Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell AlphaTec™ 04-414 Comfort PVC Gloves

Catalog No. 19014906 Shop All Ansell Products
Click to view available options
Size:
Large
This item is not returnable. View return policy
This item is not returnable. View return policy

PVC gloves offer chemical protection and secure grip performance

  • Unique rough finish with secure wet-grip performance
  • Jersey lining provide comfort and moderate insulation
  • Cut and abrasion resistant
  • 14 in./35.5 cm glove length
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 4
Certifications/Compliance 21 CFR 170-199
Certifications/Compliance (Europe) EN 420:2003 + A1:2009
Chlorinated No
Color Blue
Latex Free Yes
Material PVC, Nitrile
Size Large
EN388 Standard EN 388:3121B; EN 388:3121
EN 421 No
EN ISO 374-1 Type B
EN ISO 374-1 CHEMICAL LETTER CODES JKL
EN ISO 374 - 5 Bacteria, Fungi only
Glove Cuff Style Gauntlet
Grip Design Rough
Liner Material Jersey
Lining Lined
Model 04-414 9 211229
Series AlphaTec™ 04-414
Length (English) 14 in.
Length (Metric) 35.5 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.