Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell Alphatec™ 1800 Standard Bound Model 107 Coveralls, Hoods and Boots

Catalog No. 17999978 Shop All Ansell Products
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
This item is not returnable. View return policy
This product contains natural rubber latex which may cause allergic reactions.

Catalog No. 17-999-978

Supplier: Ansell WH18B9210708

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
This product contains natural rubber latex which may cause allergic reactions.

Coveralls with hood and boots provide entry level, low hazard liquid and particulate protection for a wide range of general industrial applications. Previously known as MICROCHEM™ by AlphaTec 1800.

  • Provides low hazard liquid spray, splash and particle protection
  • Moisture vapor permeable to help reduce the risk of heat stress
  • Low linting reduces the risk of fiber contamination in some critical areas
  • Anti-static, tested according to EN1149-5 and AATCC76 (8EMSL test method)
  • Elastic waist, wrists, and ankles
  • 2-way front zipper with storm flap
  • Bound seams, attached hood and boots
  • Available in Small through 5X-Large sizes
  • Certifications EN 1149-5, AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5, ISO 17491-4 Type 6
  • 5 years shelf life in dry, cool, covered conditions out of direct sunlight

Certifications

EN 1149-5 / AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5, ISO 17491-4 Type 6

TRUSTED_SUSTAINABILITY

Specifications

Product Type General Purpose Coveralls
Size 4X-Large
Gender Unisex
Material Microporous Film Laminate
Color White
Disposable Limited-use
Waist Style Elastic
Ankle Style Elastic, Attached Boots
Cuff Style Finger Loops
Neck Style Hooded
Hood Type 3-panel, Respirator Fit
Closure Type Front Zipper with Storm Flap
Seam Type Bound
Sterility Non-sterile
Resistance Non-toxic Particulates, Liquid, Anti-static
Certifications/Compliance AATCC 76, ANSI/ISEA 101
Certifications/Compliance (Europe) EN 1149-5, EN ISO 13982-1 Type 5, ISO 17491-4 Type 6
Description Attached Anti-skid Sure Step Sole Boots
Latex Free No
Model WH18-B-92-107
No. of Pockets None
For Use With (Application) General Maintenance, Paint Spraying, Powder Handling, Food
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.