Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell AlphaTec™ 2000 Standard Coveralls with Hood and Boots, Model 107

Catalog No. p-7070775 Shop All Ansell Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-999-869 Medium
17-999-868 Small
17-999-870 Large
17-999-871 X-Large
17-999-872 2X-Large
17-999-873 3X-Large
17-999-874 4X-Large
17-999-875 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 17-999-869 Supplier Ansell Supplier No. WH20B9210703
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Coveralls with hood and anti-skid boots feature breathable microporous laminate technology. Previously known as MICROCHEM™ by AlphaTec 2000 Standard.

  • Liquid penetration resistance and barrier to fine particulates
  • Compliant with ASTM F1761 and EN 14126:2003 resistance to penetration of blood, body fluids and blood-borne pathogens
  • Moisture vapor permeable to help reduce the risk of heat stress
  • Low linting reduces the risk of fiber contamination in some critical areas
  • Anti-static properties, tested according to EN1149-5 and AATCC76 (8EMSL test method)
  • Elastic wrists and ankles
  • 2-way front zipper with storm flap
  • Bound seams, finger loops, 3 piece respirator fit hood
  • Attached anti-skid boots with Sure Step soles
  • Applications include pharmaceutical industries, agriculture, cleanrooms, paint spraying, crime scene investigations, veterinary services, food
  • Available in small through 5X-large sizes
  • EN 14126 / ASTM F1671, DIN 32781, EN 1073-2, EN 1149-5 / AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5B, ISO 17491-4 Type 6B
  • 5 years shelf life in dry, cool, covered conditions out of direct sunlight
TRUSTED_SUSTAINABILITY

Specifications

Product Type General Purpose Coveralls
Size Medium
Gender Unisex
Material Microporous Film Laminate
Color White
Disposable Limited-use
Waist Style Elastic
Ankle Style Elastic, Attached Boots
Cuff Style Elastic, Finger Loops
Neck Style Hooded
Hood Type 3-panel, Respirator Fit
Closure Type Front Zipper with Storm Flap
Seam Type Bound
Sterility Non-sterile
Resistance Particle, Liquid, Chemicals, Anti-static
Certifications/Compliance ASTM F1761, AATCC76, ANSI/ISEA 101
Certifications/Compliance (Europe) EN 14126:2003, EN1149-5 , DIN 32781, EN 1073-2, EN ISO 13982-1 Type 5B, ISO 17491-4 Type 6B
Description Attached Sure Step sole anti-skid boots
Latex Free Yes
Model WH20-B-92-107
For Use With (Application) Life Sciences, Automotive, Metal Fabrication, Food Processing
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.