Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ansell™ Alphatec™ MICROCHEM™ 68-5000 Orange Coveralls with Hood
Shop All Ansell Products
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Description
- Protection: Barrier to numerous organic and inorganic chemicals and biological hazards; >480 minutes breakthrough time against 14 of 15 chemicals listed in EN ISO 6529
- Comfort: Multi-layer material which is lightweight, yet strong and durable
- Highly visible: Bright orange color for improved worker safety
- Design: Elastic waist, wrists and ankles, double zip closure, double cuffs with knitted inner cuff, ultrasonically welded and taped seams, 2 piece hood
- Applications: Industrial and tank cleaning, sewage purification installations, chemicals, oil and petrochemicals, pharmaceuticals, mining, agriculture, HAZMAT emergency reponse (Level B), first response and fire service
- Sizes: Available in Small through 5X-Large
- Certifications: EN 1073-2, EN 1149-5 / AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5-B, ISO 17491-4 Type 6-B, EN 14126 / ASTM F1671, EN 14605 Type 3-B, EN 14605 Type 4-B
- Storage: 5 years shelf life in dry, cool, covered conditions out of direct sunlight
Certifications
EN 1073-2, EN 1149-5 / AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5-B, ISO 17491-4 Type 6-B, EN 14126 / ASTM F1671, EN 14605 Type 3-B, EN 14605 Type 4-B

Specifications
Specifications
Size | Large |
Material | Multi-layer Barrier Laminate Fabric |
Color | Orange |
Waist Style | Elastic |
Ankle Style | Elastic |
Cuff Style | Elastic, Knit |
Neck Style | Hooded |
Hood Type | 2-panel |
Closure Type | Double Zipper |
Seam Type | Welded, Taped |
Show More |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction