Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell™ Alphatec™ MICROCHEM™ 68-6000 Gastight Coveralls with Dual Glove Locking Cuff, external link

Catalog No. p-7070766 Shop All Ansell Products
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Provide protection for emergency responders or chemical workers against dangerous and toxic chemicals in liquid or gaseous form. Ansell™ Alphatec™ MICROCHEM™ 68-6000 Gastight Coveralls with Dual Glove Locking Cuffs are Level A suits where a self-contained breathing apparatus is worn on the inside.

  • Protection:  Level A Gas tight suit tested against numerous chemicals including the ASTM F1001 test battery
  • Fabric technology:  Co-extruded multi-layer performance barrier laminate with scrim reinforcement
  • Visor technology:  Semi-rigid 3 layer PET visor providing excellent optical clarity
  • Design:  DYNAT/YKK gas-tight zipper, interchangeable locking cuff for dual glove system (external link), ultrasonically welded and taped seams
  • Applications:  Chemicals, pharmaceuticals, emergency service, sewage treatment, nuclear, petrochemicals, first response, industrial and tank cleaning, shipping
  • Sizes:  Available in Small through 3X-Large
  • Certifications:  EN 1073-2, EN1149-5 / AATCC 76, EN 14126 / ASTM F1671, EN943-1 Type 1a, EN943-2 Type 1a-ET
  • Storage:  5 years shelf life in dry, cool, covered conditions out of direct sunlight

Certifications

EN 1073-2, EN1149-5 / AATCC 76, EN 14126 / ASTM F1671, EN943-1 Type 1a, EN943-2 Type 1a-ET

TRUSTED_SUSTAINABILITY

Specifications

Color Orange
Material Barrier Laminate Fabric
Size Small
Ankle Style Attached socks, Boot overflaps
Closure Type Gas-tight zipper
Cuff Style Locking glove ring
Description Interchangeable locking cuff for dual glove system (Ansell Barrier and Neoprene), outer link
Protection Class Gas tight chemical and biological protection
Seam Type Welded, Taped

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.