Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALPPL2 Antibody (29A5), Novus Biologicals™
SDP

Catalog No. NB297957 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB297957 100 μL
NB297956 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB297957 Supplier Novus Biologicals Supplier No. NBP326510100UL
Add to Cart
Add to Cart

Human Monoclonal Antibody

ALPPL2 Monoclonal antibody specifically detects ALPPL2 in Human samples. It is validated for ELISA

Specifications

Antigen ALPPL2
Applications ELISA
Classification Monoclonal
Clone 29A5
Conjugate Unconjugated
Dilution ELISA
Formulation 50% Glycerol, 0.01M PBS, PH 7.4
Gene Alias Alkaline phosphatase Nagao isozyme, alkaline phosphatase, placental-like, alkaline phosphatase, placental-like 2, ALP-1, ALPG, ALPPL, EC 3.1.3.1, GCAP, Germ cell alkaline phosphatase, Nagao isozyme, Placental alkaline phosphatase-like, placental-like alkaline phosphatase, PLAP-like, testicular and thymus alkaline phosphatase
Host Species Human
Immunogen Recombinant Human ALPPL2 protein [UniProt P10696]
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Protein Phosphatase, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 251
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.