Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ALPPL2 Antibody (29A5), Novus Biologicals™

Human Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | ALPPL2 |
---|---|
Clone | 29A5 |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Description
ALPPL2 Monoclonal antibody specifically detects ALPPL2 in Human samples. It is validated for ELISASpecifications
ALPPL2 | |
ELISA | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Alkaline phosphatase Nagao isozyme, alkaline phosphatase, placental-like, alkaline phosphatase, placental-like 2, ALP-1, ALPG, ALPPL, EC 3.1.3.1, GCAP, Germ cell alkaline phosphatase, Nagao isozyme, Placental alkaline phosphatase-like, placental-like alkaline phosphatase, PLAP-like, testicular and thymus alkaline phosphatase | |
Recombinant Human ALPPL2 protein [UniProt P10696] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
29A5 | |
ELISA | |
Unconjugated | |
Human | |
Protein Phosphatase, Signal Transduction, Stem Cell Markers | |
50% Glycerol, 0.01M PBS, PH 7.4 | |
251 | |
IgG1 | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title