Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ALPPL2 Monoclonal Antibody (3C5A7)
GREENER_CHOICE

Catalog No. FERMA548369
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERMA548369 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERMA548369 Supplier Invitrogen™ Supplier No. MA548369
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ALPPL2 Monoclonal Antibody for Western Blot, IHC (P), Flow, ELISA

There are at least four distinct but related alkaline phosphatases: intestinal, placental, placental-like, and liver/bone/kidney (tissue non-specific). The product of this gene is a membrane bound glycosylated enzyme, localized to testis, thymus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALPPL2
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 3C5A7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene ALPG
Gene Accession No. P10696
Gene Alias Akp5; alkaline phosphatase 5; Alkaline phosphatase Nagao isozyme; Alkaline phosphatase, germ cell type; alkaline phosphatase, placental like 2; alkaline phosphatase, placental-like; alkaline phosphatase, placental-like 2; ALP-1; Alpg; ALPPL; ALPPL2; C77216; D1Ertd816e; EAP; Embryonic alkaline phosphatase; embryonic-type alkaline phosphatase; GCAP; Germ cell alkaline phosphatase; Nagao isozyme; Placental alkaline phosphatase-like; placental-like alkaline phosphatase; PLAP-like; RGD1565100; testicular and thymus alkaline phosphatase
Gene Symbols ALPG, ALPPL2
Host Species Mouse
Immunogen Purified recombinant fragment of human ALPG (AA: 170-285) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 251
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.