Learn More
Thermo Scientific™ ALPS Heat Sealer Accessories
GREENER_CHOICE
Catalog No. p-4531035
Change view
Click to view available options
Description:
384-well PCR Plate Carrier
96-Well PCR Plate Carrier
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
AB-05631000 96-Well PCR Plate Carrier
AB-0724 384-well PCR Plate Carrier
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AB-05631000 Supplier Thermo Scientific™ Supplier No. AB0563/1000
Only null left
Add to Cart
Add to Cart
Use these ALPS heat sealer accessories with the ALPS 25 Manual Heat Sealer and ALPS 50 V Microplate Heat Sealer.
Use Thermo Scientific™ ALPS heat sealer accessories with the ALPS 25 Manual Heat Sealer and ALPS 50 V Microplate Heat Sealer. The foil stripper, AB-0592, is also compatible with the ALPS 3000 Microplate Heat Sealer.
Warranty: 1 year
TRUSTED_SUSTAINABILITY
Specifications
Description 96-Well PCR Plate Carrier
For Use With (Equipment) ALPS™ Heat Sealer
No. per Pack 1 Ea.
Product Type Carrier