Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALS2CR12 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB327729 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB327729 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB327729 Supplier Novus Biologicals Supplier No. NBP305958MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ALS2CR12 Polyclonal antibody specifically detects ALS2CR12 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen ALS2CR12
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 12
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human ALS2CR12 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 130540
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.