Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALS2CR2 Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB345154 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB345154 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB345154 Supplier Novus Biologicals Supplier No. NB100442MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ALS2CR2 Polyclonal specifically detects ALS2CR2 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen ALS2CR2
Applications Western Blot
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias ALS2CR2candidate 2, Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 2 protein, CALS-21MGC102916, ILP-interacting protein, ILPIPSTRAD beta, Pseudokinase ALS2CR2, STE20-related kinase adaptor beta
Host Species Rabbit
Immunogen Residues 399-418 (SPWSELEFQFPDDKDPVWEF) of the mouse PAPK protein
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 55437
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.