Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALS2CR2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP224680SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224680SS 0.025 mg
NBP224680 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224680SS Supplier Novus Biologicals Supplier No. NBP224680SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ALS2CR2 Polyclonal specifically detects ALS2CR2 in Human, Bovine, Canine, Equine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen ALS2CR2
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2.0-4.0 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml
Formulation 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. Q9C0K7
Gene Alias ALS2CR2candidate 2, Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 2 protein, CALS-21MGC102916, ILP-interacting protein, ILPIPSTRAD beta, Pseudokinase ALS2CR2, STE20-related kinase adaptor beta
Gene Symbols STRADB
Host Species Rabbit
Immunogen A portion of amino acids 200-250 of human Als2Cr2 was used as the immunogen for the antibody.
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Apoptosis, Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 55437
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, cow, monkey and dog, 94% homologous in opossum, chicken, horse, rat, mouse and 90% homologous in zebrafish.
Target Species Human, Bovine, Canine, Equine, Primate
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.