Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALS2CR2 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$181.90 - $402.00

Specifications

Antigen ALS2CR2
Concentration 0.5 mg/mL
Dilution Western Blot 2.0-4.0 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP224680SS
SDP
View Documents
Novus Biologicals
NBP224680SS
0.025 mg
Each for $181.90
Only null left
Add to Cart
 
NBP224680
SDP
View Documents
Novus Biologicals
NBP224680
0.1 mg
Each for $402.00
Only null left
Add to Cart
 
Description

Description

ALS2CR2 Polyclonal specifically detects ALS2CR2 in Human, Bovine, Canine, Equine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

ALS2CR2
Western Blot 2.0-4.0 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml
Polyclonal
Purified
RUO
Human, Bovine, Canine, Equine, Primate
Q9C0K7
55437
A portion of amino acids 200-250 of human Als2Cr2 was used as the immunogen for the antibody.
Primary
Store at -20C. Avoid freeze-thaw cycles.
0.5 mg/mL
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Apoptosis, Protein Kinase, Signal Transduction
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
ALS2CR2candidate 2, Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 2 protein, CALS-21MGC102916, ILP-interacting protein, ILPIPSTRAD beta, Pseudokinase ALS2CR2, STE20-related kinase adaptor beta
STRADB
IgG
Protein A purified
The amino acid sequence used as immunogen is 100% homologous in human, cow, monkey and dog, 94% homologous in opossum, chicken, horse, rat, mouse and 90% homologous in zebrafish.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.