Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALS2CR3 Antibody, Novus Biologicals™
SDP

Catalog No. NBP215370 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP215370 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP215370 Supplier Novus Biologicals Supplier No. NBP215370
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ALS2CR3 Polyclonal specifically detects ALS2CR3 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen ALS2CR3
Applications Western Blot
Classification Polyclonal
Concentration 0.73mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000
Formulation 0.1M Tris-Glycine (pH7), 20% Glycerol with 0.01% Thimerosal
Gene Accession No. O60296
Gene Alias ALS2CR3GRIF1, CALS-C, GRIF-1, KIAA0549amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 3, OIP98Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 3 protein, trafficking kinesin-binding protein 2, trafficking protein, kinesin binding 2
Gene Symbols TRAK2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human TRAK2. The exact sequence is proprietary.
Molecular Weight of Antigen 101 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 66008
Test Specificity Human, Mouse. Expected to cross react based on sequence identity: Rat (95%), Bovine (96%).
Target Species Human, Mouse, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.