Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALS2CR3 Antibody (S390-43), DyLight 405, Novus Biologicals™
SDP

Catalog No. NBP259333V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP259333V 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP259333V Supplier Novus Biologicals Supplier No. NBP259333V

Mouse Monoclonal Antibody

ALS2CR3 Monoclonal specifically detects ALS2CR3 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen ALS2CR3
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone S390-43
Conjugate DyLight 405
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias ALS2CR3GRIF1, CALS-C, GRIF-1, KIAA0549amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 3, OIP98Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 3 protein, trafficking kinesin-binding protein 2, trafficking protein, kinesin binding 2
Gene Symbols TRAK2
Host Species Mouse
Immunogen Fusion protein amino acids 757-914 (C-terminus) of human TRAK2
Purification Method Protein G purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 66008
Test Specificity Detects 100kDa. Does not cross-react with TRAK1.
Target Species Human, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.