Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aluminium isopropoxide, 99.99%, (trace metal basis), ACROS Organics™
Click to view available options
Quantity:
10g
50g
250g
Packaging:
Glass Bottle
Description
For ultra trace analysis, Lewis acid
Specifications
Total Trace Metal Impurities | 150ppm max |
Boiling Point | 125.0°C to 130.0°C (38.0 mmHg) |
Color | White |
Melting Point | 129.0°C to 136.0°C |
Packaging | Glass Bottle |
Infrared Spectrum | Authentic |
Quantity | 250g |
Assay Percent Range | 99.99% |
Linear Formula | [(CH3)2CHO]3Al |
Fieser | 01,35; 03,10; 04,15; 05,14; 06,19; 08,15; 09,14; 11,29; 13,265; 15,16 |
Show More |
Safety and Handling
GHS H Statement
Flammable solid.
Causes serious eye irritation.
May cause drowsiness or dizziness.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breathing d
GHS Signal Word: Danger
EINECSNumber : 209-090-8