Aluminum chloride, anhydrous, 99.985% (metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 8848806

Quantity 5g

 View more versions of this product

Catalog No. AA8848806

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7446-70-0
    Physical Form Powder
    Assay Percent Range 99.985% (Metal basis)
    Molecular Formula AlCl3
    Formula Weight 133.34
    MDL Number MFCD00003422
    Chemical Name or Material Aluminum chloride

    Specifications

    Formula Weight 133.34
    Quantity 5g
    Color Yellow to gray
    DOT Information DOT Class: 8, Packaging Group: II
    Density 2.44g/cm3
    EINECS Number 231-208-1
    Physical Form Powder
    Odor Pungent
    Molecular Formula AlCl3
    Appearance Yellow to gray
    MDL Number MFCD00003422
    CAS 7446-70-0
    Assay Percent Range 99.985% (Metal basis)
    Chemical Name or Material Aluminum chloride

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit