Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum di(sec-butoxide)acetoacetic ester chelate, tech., Al 8.4% min

Catalog No. aa8934936
Click to view available options
Quantity:
500 g

Catalog No. AA8934936

Supplier: Thermo Scientific Chemicals 08934936

Only null left
Add to Cart

CAS: 24772-51-8 | C14H27AlO5

Aluminum di(sec-butoxide)acetoacetic ester chelate is used in addition with lithium acetate and titanium(IV) ethoxide to prepare precursor sol to prepare Li1.3Al0.3Ti1.7(PO4)3 thin films on glass substrate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24772-51-8
Molecular Formula C14H27AlO5
MDL Number MFCD00084762

Specifications

Density 1.03 g/mL
Flash Point 76°C (169°F)
Odor Alcohol-like
Quantity 500 g
Sensitivity Moisture sensitive
Solubility Information Immiscible with water.
Formula Weight 302.3
Grade Technical
Physical Form Liquid
Chemical Name or Material Aluminum di (sec-butoxide)acetoacetic ester chelate
Hazard Category H227-H315-H319
Hazard Statement GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Precautionary Statement P210-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c
EINECSNumber 246-455-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.