Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum lactate, 90-95%, contains ^=5% water

Catalog No. AA3912236
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3912236 500 g
AA3912222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3912236 Supplier Thermo Scientific Chemicals Supplier No. 03912236
Only null left
Add to Cart

CAS: 18917-91-4 | C9H15AlO9 | 294.19 g/mol

It is used as a skin moisturizer lotion to treat dry, scaly, itchy skin, reagent in biological activity studies of aluminum and in sol-gel processing. Also used in fire foam. Aluminum L-Lactate has been used to inject aluminum into rabbits. Aluminum L-Lactate has been shown to decrease human enamel dissolution in a pH 4 acetate buffer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18917-91-4
Molecular Formula C9H15AlO9
Molecular Weight (g/mol) 294.19
MDL Number MFCD20529175
InChI Key VXYADVIJALMOEQ-UHFFFAOYNA-K
IUPAC Name bis[(2-hydroxypropanoyl)oxy]alumanyl 2-hydroxypropanoate
SMILES CC(O)C(=O)O[Al](OC(=O)C(C)O)OC(=O)C(C)O

Specifications

Physical Form Powder
Quantity 500 g
Merck Index 14,348
Solubility Information Very soluble in cold water
Formula Weight 294.2
Percent Purity 90 to 95%
Concentration or Composition (by Analyte or Components) Contains ≈5% water
Odor Odorless
Chemical Name or Material Aluminum lactate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 242-670-9
RTECSNumber BD2214000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.