Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aluminum oxalate hydrate, 99%
Click to view available options
Quantity:
10 g
50 g
250 g
1 kg
Description
Aluminum oxalate is used as a mordant for printing textiles and dyeing cotton. It finds use as a laboratory reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | White |
| Linear Formula | Al2(C2O4)3·xH2O |
| Quantity | 10 g |
| Merck Index | 14,355 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 318.02 (Anhydrous) |
| Percent Purity | 99% |
| Physical Form | Chunks and/or Powder |
| Chemical Name or Material | Aluminum oxalate hydrate |
Safety and Handling
| Hazard Category | H302+H312 |
| Hazard Statement | GHS H Statement H302-H312 Harmful if swallowed. Harmful in contact with skin. |
| Precautionary Statement | P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c |
| EINECSNumber | 212-407-2 |
| RTECSNumber | BD0516250 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction