Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum phosphate, 97%, Alfa Aesar™

Manufacturer:  Alfa Aesar 1110930

Quantity 250g

 View more versions of this product

Catalog No. AA1110930


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7784-30-7
97%
121.95
Aluminum phosphate
Powder
AlO4P
MFCD00003430

Specifications

7784-30-7
97%
White powder
MFCD00003430
White
Aluminum phosphate
Powder
AlO4P
121.95
2.566g/cm3
250g

Safety and Handling

SDS & Safety Documents

H314-H318

DOTInformation : DOT Class: 8, Packaging Group: III

EINECSNumber : 232-056-9

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit