Aluminum phosphate, 99.99% (metals basis), Alfa Aesar™, Puratronic™

Manufacturer: Alfa Aesar 1062814

Quantity 25g

 View more versions of this product

Catalog No. AA1062814

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7784-30-7
    Physical Form Powder
    Assay Percent Range 99.99% (Metal basis)
    Molecular Formula AlO4P
    Formula Weight 121.95
    MDL Number MFCD00003430
    Chemical Name or Material Aluminum phosphate

    Specifications

    Formula Weight 121.95
    Quantity 25g
    Color White
    DOT Information DOT Class: 8, Packaging Group: III
    EINECS Number 232-056-9
    Physical Form Powder
    Assay Percent Range 99.99% (Metal basis)
    Molecular Formula AlO4P
    Appearance White powder
    MDL Number MFCD00003430
    CAS 7784-30-7
    Health Hazard 1 H314-H318
    Chemical Name or Material Aluminum phosphate

    H314-H318

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit