Learn More
Aluminum phosphate, Puratronic™, 99.99% (metals basis)
Description
Aluminum phosphate is used in the production of glass, mixture of ceramic, waterproofing concrete and dental cements, in cosmetics as a emollient, flame retardant, catalyst in organic synthesis, fireproofing textile, pharmaceuticals and dyes. It also used in many adhesives, binders and cements.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | >1500°C |
Color | White |
Physical Form | Powder |
Quantity | 5 g |
Assay Percent Range | 99.99% (Metal basis) |
Solubility Information | Soluble in concentrated hydrochloric acid,alcohol and nitric acid. Insoluble in water. |
Formula Weight | 121.95 |
Appearance | White powder |
Density | 2.566 g/cm3 |
Chemical Name or Material | Aluminum phosphate |
Safety and Handling
Hazard Category | H315-H320 |
Precautionary Statement | P264b-P280-P302+P352-P305+P351+P338-P315-P362-P501c |
DOTInformation | DOT Class: 8, Packaging Group: III |
EINECSNumber | 232-056-9 |
RTECSNumber | TB6450000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.