Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum rod, 6mm (0.2in) dia, Puratronic™, 99.9965% (metals basis)

Catalog No. p-4781780
Click to view available options
Quantity:
15 cm
5 x 15 cm

Catalog No. AA11478BT

Supplier: Thermo Scientific Chemicals 011478BT

Only null left
Add to Cart

Form: Approx. 0.76g/cm | CAS: 7429-90-5 | Al | 26.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7429-90-5
Molecular Formula Al
Molecular Weight (g/mol) 26.98
MDL Number MFCD00134029
InChI Key XAGFODPZIPBFFR-UHFFFAOYSA-N
Synonym aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
PubChem CID 5359268
ChEBI CHEBI:33629
IUPAC Name aluminium
SMILES [Al]

Specifications

Color Silver
Appearance Silver-colored rod
Density 2.7g/cm3
Quantity 15 cm
Odor Odorless
Form Rod
Assay Percent Range 99.9965% (Metals basis)
Chemical Name or Material Aluminum
EINECSNumber 231-072-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.