AM 251, Tocris

Potent CB1 antagonist; Also GPR55 agonist

Manufacturer: R&D Systems 1117/1

Quantity 1mg

 View more versions of this product

Catalog No. 11-171

  Check Availability
Add to Cart

    Chemical Identifiers

    Assay Percent Range >99%
    Molecular Formula C22H21Cl2IN4O
    Formula Weight 555.24
    Synonym N-(Piperidin-1-yl)-5-(4-iodophenyl)-1-(2, 4-dichlorophenyl)-4-methyl-1H-pyrazole-3-carboxamide
    Chemical Name or Material AM 251

    Specifications

    Formula Weight 555.24
    Quantity 1mg
    Assay Percent Range >99%
    Molecular Formula C22H21Cl2IN4O
    CAS 183232-66-8
    Synonym N-(Piperidin-1-yl)-5-(4-iodophenyl)-1-(2, 4-dichlorophenyl)-4-methyl-1H-pyrazole-3-carboxamide
    Recommended Storage Store at Room Temperature
    Chemical Name or Material AM 251

    Store at Room Temperature

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit