Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ AM 4113
GREENER_CHOICE

Catalog No. p-7114674
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective, high affinity CB1 antagonist

AM 4113 is a high affinity and selective CB1 antagonist (Kd = 0.89 nM). Exhibits 100-fold selectivity for CB1 over CB2. Attenuates CB1 agonist AM 411-induced locomotor suppression and reduces food intake in vivo, but does not induce signs of nausea.

Specifications

Chemical Name or Material 5-(4-Chlorophenyl)-1-(2,4-dichlorophenyl)-4-methyl-1H-pyrazole-3-carboxamide
CAS 614726-85-1
Assay Percent Range >98%
Quantity 10 mg
Molecular Formula C17H12Cl3N3O
Formula Weight Observed MW: 380.66
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.