Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Amber Reagent Bottle , Narrow Mouth with Acid Proof Polypropylene Stopper
SCI_ED

Catalog No. S27977 Shop All Eisco Products
Click to view available options
Capacity (Metric):
30 mL
60 mL
125 mL
250 mL
500 mL
1000 mL
2000 mL

Catalog No. S27977

Supplier: Eisco™ CH0161E

Only null left
Add to Cart
Add to Cart

Narrow Mouth with Acid Proof Polypropylene Stopper. Made of high quality borosilicate glass.

Specifications

Capacity 500 mL
Capacity (Metric) 500 mL
Capacity (English) 17 oz.
Color Amber
Description Socket Size: 24/29
For Use With (Application) To contain chemicals in liquid or powder form
Material Borosilicate Glass
Length (Metric) 22.86 cm
Quantity 1
Sterility Non-sterile
Standard Taper Joint 24/29
Weight (Metric) 362.87 g
Width (Metric) 228.6 mm
Closure Color White
Closure Included Yes
Disposable Reusable
Graduated No
Mouth Narrow
Shape Cylindrical
Height (Metric) 12.7 cm
Product Type Reagent Bottle
For Use With (Equipment) Acid
Height (English) 5 in.
Chemical Label Unlabeled
Closure Material Polypropylene
Closure Size 24/29
Closure Type Stopper
Autoclavable Autoclavable
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.