Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amberjet™ 4200-Cl

Catalog No. AC301330025
Change view
Click to view available options
Quantity:
2.5 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC301330025 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC301330025 Supplier Thermo Scientific Chemicals Supplier No. 301330025

CAS: 60177-39-1 | C22H31ClN | 344.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60177-39-1
Molecular Formula C22H31ClN
Molecular Weight (g/mol) 344.95
MDL Number MFCD00145586

Specifications

Physical Form Beads
Particle Size 0.600 to 0.800mm harmonic mean size, 0.5% Max. (<0.425mm), 5.0% Max. (>0.850mm), 1.25 uniformity coefficient Max.
Volume Total Capacity (Cl): 1.30 eq/L min.
Water Retention Capacity (Cl): 49% to 55%
Color Yellow
Quantity 2.5 kg
Chemical Name or Material Amberjet™ 4200-Cl

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.