Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amberlite™ IRN-77, ion exchange resin, nuclear grade

Catalog No. p-4408669
Click to view available options
Quantity:
500 g
2 kg

Catalog No. AA1682136

Supplier: Thermo Scientific Chemicals 01682136

Only null left
Add to Cart

Styrene-DVB; Strongly acidic cation exchange resin | CAS: 11128-94-2 | Styrene-DVB

Amberlite™ IRN-77 is used in the primary water treatment for the removal of fission products, activated corrosion products and suspended matter. It is also used to control the pH of the reactor coolant stream by removing the excess lithium. Further, it is used in the removal of radioactive cations such as Cesium from waste streams. In addition to this, it is used as a cation exchanger to determine the sorption activity towards cesium(I) and strontium(II) using isothermal titration calorimetry and solution-depletion method.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 11128-94-2
Molecular Formula Styrene-DVB
MDL Number MFCD00145822

Specifications

Physical Form Beads
Odor Odorless
Color Brown
Quantity 500 g
Solubility Information Insoluble in water,acid and alkali and various organic solvents.
Grade Nuclear
Chemical Name or Material Amberlite IRN-77
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.