Learn More
Invitrogen™ CellsDirect™ One-Step qRT-PCR Kit
Description
Includes
10mL Resuspension Buffer; 1mL Lysis Enhancer; 500μL DNase I, Amplification Grade (1U/μL); 160μL 10X DNase I Buffer; 400μL 25mM EDTA; 100μL SuperScript III RT/Platinum Taq Mix; 2 x 1.25mL 2X Reaction Mix; 1mL 50mM MgSO4; 2mL DEPC-treated water; 10μL HeLa Total RNA (10ng/μL); 100μL ROX Reference Dye
The CellsDirect One-Step qRT-PCR Kit delivers highly sensitive and specific real-time qPCR results directly from cells, without the need for an RNA purification step.
• Efficiency: offers significant time and cost savings for increased throughput
• Convenience: enzyme mix includes both SuperScript™ III Reverse Transcriptase and Platinum™ Taq DNA Polymerase
• Sensitivity: no separate RNA isolation step increases sensitivity by minimizing sample loss
• Broad detection range: detect targets from samples ranging from a single cell to 10,000 cells
• Versatility: compatible with a wide variety of cell lines and samples collected by Laser Capture Microdissection (LCM)
Streamline your workflow
In traditional real-time quantitative RT-PCR (qRT-PCR), you first isolate RNA from cells in a time-consuming procedure that can lead to loss of material. Using the CellsDirect One-Step qRT-PCR Kit, you lyse the cells and add the complete lysate directly to a one-step qRT-PCR reaction with minimal handling and sample loss.
Ideal for your qRT-PCR application
CellsDirect technology is validated for gene expression studies involving limited samples. CellsDirect One-Step qRT-PCR Kits are especially valuable for high-throughput applications by eliminating costly, time-consuming RNA purification steps. The CellsDirect One-Step qRT-PCR Kit may also be used to detect genomic DNA targets. CellsDirect One-Step qRT-PCR Kits may be used in conjunction with hydrolysis probes (e.g., TaqMan™ or LUX™ fluorogenic primers).
This kit includes a separate tube of ROX™ Reference Dye. An equivalent kit with ROX Reference Dye included in the 2X Reaction Mix is also available.
Order Info
Shipping Condition: Dry ice
Specifications
Specifications
| Concentration | 2X |
| Content And Storage | • 10 mL Resuspension Buffer • 1 mL Lysis Enhancer • 500 μL DNase I, Amplification Grade (1 U/μL) • 160 μL 10X DNase I Buffer • 400 μL 25 mM EDTA • 100 μL SuperScript™ III RT/Platinum™ Taq Mix • 2 × 1.25 mL 2X Reaction Mix • 1 mL 50 mM MgSO4 • 2 mL DEPC-treated water • 10 μL HeLa Total RNA (10 ng/μL) • 100 μL ROX™ Reference Dye (store at -20°C in the dark) The kit is shipped on dry ice and should be stored at -20°C |
| Detection Method | Primer-probe |
| Reaction Speed | Fast or Standard |
| For Use With (Equipment) | 7500 Fast System, 7500 System, BioRad iQ5, BioRad MyiQ, Corbett RotorGene, MJ Opticon, Stratagene Mx3000P, Stratagene Mx3005P, Stratagene Mx4000, BioRad iCycler iQ, Cepheid SmartCycler, MJ Chromo4, QuantStudio 3 and 5 systems, QuantStudio 6 and 7 Pro systems, QuantStudio 7 Flex system, QuantStudio 12k Flex system, QuantStudio 6 Flex system |
| Product Line | CellsDirect, Platinum, SuperScript |
| Quantity | 100 reactions |
| For Use With (Application) | RT-PCR |
| No. of Reactions | 100 reactions |
| Sample Type | Cells |
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.