Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Gel Loading Buffer II (Denaturing PAGE)

Catalog No. p-4926900
Encompass_Preferred
Click to view available options
Quantity:
1 mL
10 mL

A 1–2X solution of 95% Formamide, 18 mM EDTA, and 0.025% SDS, Xylene Cyanol, and Bromophenol Blue.

A 1–2X solution of 95% Formamide, 18 mM EDTA, and 0.025% SDS, Xylene Cyanol, and Bromophenol Blue. Supplied in one 1.4 mL bottle. All Invitrogen™ Gel Loading Solutions are rigorously tested for non-specific endonuclease activity, exonuclease activity, RNase activity, and for functionality. These are the same solutions we use in-house and in our kits.

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at –20°C.
Buffer Drying Solutions
Format Bottle
Label or Dye Xylene Cyanol
Form Liquid
Gel Compatibility Denaturing Polyacrylamide Gels
Product Line Ambion™
Quantity 1 mL
Shipping Condition Room Temperature
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.