Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Proteinase K Powder

Catalog No. p-4926916
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg

Proteinase K from the fungus Engyodontium album is a non-specific serine protease that is useful for general digestion of proteins.

Proteinase K from the fungus Engyodontium album is a non-specific serine protease that is useful for general digestion of proteins.

Proteinase K Remains Active:
• Over a wide pH range (optimal activity between 6.5 and 9.5)
• Under denaturing conditions (e.g. the presence of SDS or urea)
• In the presence of metal chelating agents (e.g. EDTA)
• At comparatively high temperatures (optimum digestion temperature is 65°C)

Applications: Removal of endogenous nucleases during the preparation of DNA and RNA; preparation of tissue sections for in situ hybridization.

Performance and Quality Testing: Endodeoxyribonuclease and exodeoxyribonuclease assays; liquid form tested for absence of RNase activity.

Unit Definition: One mAnson unit is described as that amount of enzyme that liberates 1 μmole of Folin-positive amino acid within one minute at 37°C using hemoglobin as a substrate.

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Host Species T. album
Packaging Bottle
Content And Storage Store dry powder in desiccant at –20°C.
Shipping Condition Dry Ice
Quantity 100 mg
Product Type Proteinase K
Product Line Ambion™
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.