Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AMBRA1 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP107124 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mL
0.2 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP107124 0.2 mL
NBP107124SS 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP107124 Supplier Novus Biologicals Supplier No. NBP107124
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 3 publications

AMBRA1 Polyclonal specifically detects AMBRA1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen AMBRA1
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:400, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:400
Formulation PBS and 30% Glycerol with 0.05% Sodium Azide
Gene Accession No. A2AH22
Gene Alias activating molecule in BECN1-regulated autophagy protein 1, autophagy/beclin-1 regulator 1, DCAF3, DDB1 and CUL4 associated factor 3, FLJ20294, KIAA1736activating molecule in beclin-1-regulated autophagy, MGC33725, WD repeat domain 94, WDR94
Gene Symbols AMBRA1
Host Species Rabbit
Immunogen A synthetic peptide derived from mouse activating molecule in Beclin 1-regulated/AMBRA1 protein sequence [amino acids range 1175-1250] [UniProt # A2AH22]
Purification Method Affinity Purified
Quantity 0.2 mL
Regulatory Status RUO
Research Discipline Autophagy
Primary or Secondary Primary
Gene ID (Entrez) 55626
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.