Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AMDHD1 Antibody, Novus Biologicals™
SDP

Catalog No. NB5672120UL Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB5672120UL 20 μL
NBP156721 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB5672120UL Supplier Novus Biologicals Supplier No. NBP15672120UL

Rabbit Polyclonal Antibody

AMDHD1 Polyclonal specifically detects AMDHD1 in Human samples. It is validated for Western Blot.

Specifications

Antigen AMDHD1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q96NU7
Gene Alias amidohydrolase domain containing 1, Amidohydrolase domain-containing protein 1, EC 3.5.2.7, MGC35366, probable imidazolonepropionase
Gene Symbols AMDHD1
Host Species Rabbit
Immunogen Synthetic peptides corresponding to AMDHD1(amidohydrolase domain containing 1) The peptide sequence was selected from the N terminal of AMDHD1. Peptide sequence AVLEGASLVVGKDGFIKAIGPADVIQRQFSGETFEEIIDCSGKCILPGLV.
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 144193
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.