Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AMDHD1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | AMDHD1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB5672120UL
![]() |
Novus Biologicals
NBP15672120UL |
20 μL |
Each for $206.00
|
|
|||||
NBP156721
![]() |
Novus Biologicals
NBP156721 |
100 μL |
Each for $487.50
|
|
|||||
Description
AMDHD1 Polyclonal specifically detects AMDHD1 in Human, Mouse samples. It is validated for Western Blot.Specifications
AMDHD1 | |
Polyclonal | |
Rabbit | |
Q96NU7 | |
144193 | |
Synthetic peptides corresponding to AMDHD1(amidohydrolase domain containing 1) The peptide sequence was selected from the N terminal of AMDHD1. Peptide sequence AVLEGASLVVGKDGFIKAIGPADVIQRQFSGETFEEIIDCSGKCILPGLV. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
amidohydrolase domain containing 1, Amidohydrolase domain-containing protein 1, EC 3.5.2.7, MGC35366, probable imidazolonepropionase | |
AMDHD1 | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title