Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

American Educational Products Dropping Bottles
SCI_ED

Catalog No. p-2366841 Shop All American Educational Products LLC Products
Click to view available options
Capacity (Metric):
15 mL
30 mL
250 mL
Color:
Amber
Clear
Shape:
French Square
Round
This item is not returnable. View return policy
This item is not returnable. View return policy

Store reagent solutions in these sturdy dropping bottles.

  • Come with Bakelite™ screw tops, glass pipets and rubber bulbs
  • Available in French Square or Boston Round shape, clear or amber color and a wide range of sizes

Order Info

Replacement pipets available for each bottle (See Cat. No. S31671P).

Specifications

Capacity 0.8 oz., 15 mL
Capacity (Metric) 15 mL
Capacity (English) 0.8 oz.
Color Clear
Includes Dropper assembly
Material Glass
Shape French Square
Closure Type Screw Cap
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.