Learn More
Cytiva Amersham™ CyDye™ Fluorescent Nucleotides
Label DNA and RNA probes. Cytiva Life Sciences™ Amersham™ CyDye™ Fluorescent Nucleotides are optimized for multicolor analysis in microarrays, FISH, chromosome identification, whole chromosome painting, karyotyping, and gene mapping.
$734.54 - $5560.63
Products 10
Catalog Number Mfr. No. Product Type Quantity Color Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Quantity Color Price Quantity & Availability  
45-001-238
View Documents
Cytiva
PA53026
Cy3-UTP 100 nmol Orange
Each for $1,209.77
Only null left
Add to Cart
45-001-241
View Documents
Cytiva
PA55322
Value pack containing 5 × 25 nmol Cy3-dUTP + 5 × 25 nmol Cy5-dUTP 5 x 25 nmol Orange, Far-Red
Each for $5,560.63
Only null left
Add to Cart
45-000-738
View Documents
Cytiva
PA53022
Cy3-dUTP 25 nmol Orange
Each for $734.54
Only null left
Add to Cart
45-000-737
missing translation for 'viewDocuments'
Cytiva
PA53021
Cy3-dCTP 25 nmol Orange
Each for $734.54
Only null left
Add to Cart
45-000-739
missing translation for 'viewDocuments'
Cytiva
PA55021
Cy5-dCTP 25 nmol Far-Red
Each for $755.15
Only null left
Add to Cart
45-000-740
missing translation for 'viewDocuments'
Cytiva
PA55022
Cy5-dUTP 25 nmol Far-Red
Each for $755.15
Only null left
Add to Cart
45-500-149
missing translation for 'viewDocuments'
Cytiva
PA53521
Cy3.5-dCTP 25 nmol Scarlet
Each for $743.23
Only null left
Add to Cart
45-001-290
missing translation for 'viewDocuments'
Cytiva
25801086
Cy3-CTP 100 nm Orange N/A N/A N/A
45-001-291
missing translation for 'viewDocuments'
Cytiva
25801087
Cy5-CTP 100 nmol Far-Red N/A N/A N/A
45-001-239
missing translation for 'viewDocuments'
Cytiva
PA55026
Cy5-UTP 100 nmol Far-Red N/A N/A N/A
Description
Description
For labeling DNA and RNA probes especially for multicolor analysis in microarrays, FISH, chromosome identification, whole chromosome painting, karyotyping, and gene mapping. CyDye labeled nucleotides are available in a range of bright, intense colors with narrow emission bands—ideal for multiplexing within a single sample.
  • Low nonspecific binding
  • High photostability
  • High water solubility
  • pH insensitive
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.