Description
- Protein binding capacity over over 200 μg/cm2 for easy signal detection
- High protein retention even after harsh washing steps
- High protein capture during transfer minimizes sample loss
- Available in popular precut sizes for your application
Specifications
Specifications
| Quantity | 5/Pk. |
| Pore Size | 0.45 μm |
| Dimensions (LxW) | 300 x 600mm |