Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Amersham™ Hybond™ PVDF Membranes: Rolls

Catalog No. 10600030 Shop All Cytiva Products
Change view
Click to view available options
Dimensions (LxW):
4 m x 254 mm
4 m x 260 mm
Grade:
Hybond LFP
Hybond P
Hybond SEQ
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (LxW) Grade
10-600-030 4 m x 260 mm Hybond SEQ
45-004-021 4 m x 260 mm Hybond P
45-004-095 4 m x 254 mm Hybond LFP
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 10-600-030 Supplier Cytiva Supplier No. 10600030
Add to Cart
Add to Cart

Ideal for Western blotting applications where stripping and reprobing are necessary. Cytiva Life Sciences™ Amersham™ Hybond™ PVDF Membranes: Rolls are hydrophobic PVDF membranes.

p>Supplied in rolls.

  • Hydrophobic PVDF membranes
  • Small pore size eliminates blow-through and increases protein binding over a wide range of molecular weights

Amersham Hybond 0.2μm

  • Compatible with a range of solvents used for rapid destaining
  • Amersham Hybond SEQ 0.2μm
    • Specifically designed for protein sequencing applications and solid phase amino acid analysis
    • Offers excellent blotting for proteins Mr<20,000
    Amersham Hybond LFP 0.2μm
    • Specifically optimized for fluorescence detection in Western blotting applications
    • Provides low background fluorescence, resulting in high sensitivity

    TRUSTED_SUSTAINABILITY

    Specifications

    For Use With (Application) Specifically designed for protein sequencing applications and solid phase amino acid analysis
    Format Roll
    Pore Size 0.2 μm
    Dimensions (LxW) 4 m x 260 mm
    Grade Hybond SEQ
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.