Learn More
Description
Individual sheets.
- Hydrophobic PVDF membranes
- Small pore size eliminates blow-through and increases protein binding over a wide range of molecular weights
Amersham Hybond 0.2μm
- Compatible with a range of solvents used for rapid destaining
- Specifically designed for protein sequencing applications and solid phase amino acid analysis
- Offers excellent blotting for proteins Mr<20,000
- Specifically optimized for fluorescence detection in Western blotting applications
- Provides low background fluorescence, resulting in high sensitivity
Specifications
Specifications
| For Use With (Application) | Ideal for Western blotting applications where stripping and reprobing are necessary |
| Quantity | 25/Pk. |
| Format | Sheet |
| Material | PVDF |
| Dimensions (LxW) | 8 x 9 cm |
| Product Type | Hybond P |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.