Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Amersham™ Hybond™ PVDF Membranes: Sheets
GSA_VA

Catalog No. 45004097 Shop All Cytiva Products
Change view
Click to view available options
Dimensions (LxW):
8 x 9 cm
20 x 20 cm
Format:
Sheet
Quantity:
10/Pk.
25/Pk.
Product Type:
Hybond LFP
Hybond P
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (LxW) Format Quantity Product Type
45-004-097 20 x 20 cm Sheet 10/Pk. Hybond LFP
45-004-046 20 x 20 cm Sheet 10/Pk. Hybond P
45-004-079 8 x 9 cm Sheet 25/Pk. Hybond P
45-004-100 8 x 9 cm Sheet 25/Pk. Hybond LFP
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 45-004-097 Supplier Cytiva Supplier No. 10600060
Add to Cart
Add to Cart

Ideal for Western blotting applications where stripping and reprobing are necessary

Individual sheets.

  • Hydrophobic PVDF membranes
  • Small pore size eliminates blow-through and increases protein binding over a wide range of molecular weights

Amersham Hybond 0.2μm

  • Compatible with a range of solvents used for rapid destaining
  • Amersham Hybond SEQ 0.2μm
    • Specifically designed for protein sequencing applications and solid phase amino acid analysis
    • Offers excellent blotting for proteins Mr<20,000
    Amersham Hybond LFP 0.2μm
    • Specifically optimized for fluorescence detection in Western blotting applications
    • Provides low background fluorescence, resulting in high sensitivity

    This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

    TRUSTED_SUSTAINABILITY

    Specifications

    For Use With (Application) Ideal for Western blotting applications where stripping and reprobing are necessary
    Quantity 10/Pk.
    Format Sheet
    Material PVDF
    Dimensions (LxW) 20 x 20 cm
    Product Type Hybond LFP
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.