Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Amersham™ Hybond™ PVDF Membranes: Sheets

Catalog No. p-4696456
Click to view available options
Dimensions (LxW):
8 x 9 cm
20 x 20 cm
Format:
Sheet
Quantity:
10/Pk.
25/Pk.
Product Type:
Hybond LFP
Hybond P

Ideal for Western blotting applications where stripping and reprobing are necessary

Individual sheets.

  • Hydrophobic PVDF membranes
  • Small pore size eliminates blow-through and increases protein binding over a wide range of molecular weights

Amersham Hybond 0.2μm

  • Compatible with a range of solvents used for rapid destaining
  • Amersham Hybond SEQ 0.2μm
    • Specifically designed for protein sequencing applications and solid phase amino acid analysis
    • Offers excellent blotting for proteins Mr<20,000
    Amersham Hybond LFP 0.2μm
    • Specifically optimized for fluorescence detection in Western blotting applications
    • Provides low background fluorescence, resulting in high sensitivity

    TRUSTED_SUSTAINABILITY

    Specifications

    For Use With (Application) Ideal for Western blotting applications where stripping and reprobing are necessary
    Quantity 10/Pk.
    Format Sheet
    Material PVDF
    Dimensions (LxW) 20 x 20 cm
    Product Type Hybond P
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Your feedback has been submitted: Thank you for helping us improve our website.