Learn More
Cytiva Amersham™ Hypercassette™ Autoradiography Cassette
Use for the toughest demands of research in the life sciences. Cytiva Life Sciences™ Amersham™ Hypercassette™ Autoradiography Cassettes are constructed from strong aluminum and a choice of colors enables color coding of work.
Specifications
Description Amersham hypercssette autoradiography cassettes are designed to survive the toughest demands of research in the life sciences.
For Use With (Application) Light-tight exposures at temperatures below -70°C
View More Specs
Products 2
Catalog Number Mfr. No. Dimensions (LxW) Color Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (LxW) Color Price Quantity & Availability  
45-000-758
View Documents
Cytiva
RPN11649
8 x 10 in. Neutral (Standard) N/A N/A N/A
45-200-586
View Documents
Cytiva
RPN11650
10 x 12 in. Neutral (Standard) N/A N/A N/A
Description
Description
    • Robust and chip-proof autoradiography cassettes.
    • Light-tight exposures at temperatures as low as -70°C.
    • Maximum contact between film and sample provides optimal resolution.
    • Nonporous pad gives a wipe-clean surface ensuring minimum contamination
Specifications
Specifications
Amersham hypercssette autoradiography cassettes are designed to survive the toughest demands of research in the life sciences.
Light-tight exposures at temperatures below -70°C
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.