Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytiva Amersham™ Hyperfilm™ ECL
Click to view available options
Dimensions (LxW):
12.7 x 17.8 cm (5 x 7 in.)
18 x 24 cm (7.1 x 9.5 in.)
20.3 x 25.4 cm (8 x 10 in.)
Quantity:
100 Sheets/Box
50 Sheets/Box
Description
- Antistatic coating reduces risk of visible artifacts
- Clear base offers high contrast for easy reading of images
- Suitable for use with all blue and green light chemiluminescent systems
- Film can be processed in automatic processors or manually using most common X-ray film developers and fixers
- Chemiluminescent detection in Western, Southern, and Northern blots

Specifications
Specifications
Product Type | Amersham Hyperfilm ECL |
For Use With (Application) | Chemiluminescent detection in Western, Southern, and Northern blots. |
Quantity | 100 Sheets/Box |
Storage Requirements | 4°C |
Dimensions (LxW) | 18 x 24 cm (7.1 x 9.5 in.) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction