Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Amersham™ Protran™ NC Nitrocellulose Membranes: Rolls

Catalog No. 45004005 Shop All Cytiva Products
Change view
Click to view available options
Width (Metric):
15 cm
20 cm
30 cm
Pore Size:
0.1 μm
0.2 μm
0.45 μm
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Width (Metric) Pore Size
45-004-005 20 cm 0.1 μm
45-004-000 30 cm 0.1 μm
45-004-002 30 cm 0.45 μm
45-004-001 30 cm 0.2 μm
45-004-006 20 cm 0.2 μm
45-004-010 15 cm 0.1 μm
45-004-012 15 cm 0.45 μm
45-004-007 20 cm 0.45 μm
45-004-011 15 cm 0.2 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 45-004-005 Supplier Cytiva Supplier No. 10600005
Add to Cart
Add to Cart

Use this versatile nitrocellulose membrane for excellent binding affinity for small proteins and peptides, as well as nucleic acids. Cytiva Amersham™ Protran™ NC Nitrocellulose Membranes: Rolls offer excellent sensitivity, resolution, and low background.

Packed in rolls.

  • 100% pure nitrocellulose
  • High affinity for proteins
  • Blocking ability
  • Compatible with a range of detection methods, including chemiluminescent, chromogenic, and fluorescent

TRUSTED_SUSTAINABILITY

Specifications

Length (Metric) 4 m
Width (Metric) 20 cm
For Use With (Application) Western blots, dot-blot assays, and other protein or nucleic acid methods
Format Roll
Material 100% Pure Nitrocellulose
Pore Size 0.1 μm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.