Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Amersham™ Protran™ NC Nitrocellulose Membranes: Sheets

Catalog No. 45004025 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
10/Pk.
25/Pk.
5/Pk.
Length (Metric):
8 cm
10 cm
10.2 cm
16 cm
20 cm
60 cm
Width (Metric):
9 cm
10 cm
13.3 cm
14 cm
20 cm
30 cm
Pore Size:
0.1 μm
0.2 μm
0.45 μm
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Length (Metric) Width (Metric) Pore Size
45-004-025 5/Pk. 60 cm 30 cm 0.45 μm
45-004-024 5/Pk. 60 cm 30 cm 0.2 μm
45-004-031 25/Pk. 20 cm 20 cm 0.45 μm
45-004-032 10/Pk. 20 cm 20 cm 0.45 μm
45-004-033 25/Pk. 20 cm 20 cm 0.2 μm
45-004-034 10/Pk. 20 cm 20 cm 0.2 μm
45-004-035 25/Pk. 20 cm 20 cm 0.1 μm
45-004-036 10/Pk. 20 cm 20 cm 0.1 μm
45-004-048 25/Pk. 16 cm 14 cm 0.45 μm
45-004-056 10/Pk. 10 cm 10 cm 0.45 μm
45-004-072 25/Pk. 8 cm 9 cm 0.45 μm
45-004-073 25/Pk. 8 cm 9 cm 0.2 μm
45-500-101 10/Pk. 10.2 cm 13.3 cm 0.2 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Catalog No. 45-004-025 Supplier Cytiva Supplier No. 10600033
Add to Cart
Add to Cart

Use for Western blots, dot-blot assays, and other protein or nucleic acid methods. Cytiva Amersham™ Protran™ NC Nitrocellulose Membranes: Sheets offer excellent sensitivity, resolution, and low background.

Individual sheets.

  • 100% pure nitrocellulose
  • High affinity for proteins
  • Blocking ability
  • Compatible with a range of detection methods, including chemiluminescent, chromogenic, and fluorescent

TRUSTED_SUSTAINABILITY

Specifications

Length (Metric) 60 cm
Width (Metric) 30 cm
For Use With (Application) Western blots, dot-blot assays, and other protein or nucleic acid methods
Quantity 5/Pk.
Format Sheet
Pore Size 0.45 μm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.