Learn More
Description
Use for Western blots, dot-blot assays, and other protein or nucleic acid methods. Cytiva Amersham™ Protran™ NC Nitrocellulose Membranes: Sheets offer excellent sensitivity, resolution, and low background.
Individual sheets.
- 100% pure nitrocellulose
- High affinity for proteins
- Blocking ability
- Compatible with a range of detection methods, including chemiluminescent, chromogenic, and fluorescent
Specifications
Specifications
| Length (Metric) | 20 cm |
| Width (Metric) | 20 cm |
| For Use With (Application) | Western blots, dot-blot assays, and other protein or nucleic acid methods |
| Quantity | 25/Pk. |
| Format | Sheet |
| Pore Size | 0.45 μm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.