Learn More
Description
Supplied in rolls.
- Low background enables a dynamic range for complicated samples
- Suitable for Western blots, dot-blot assays, and other protein or nucleic acid methods
- 100% pure nitrocellulose
- High affinity for proteins
- Blocking ability
Specifications
Specifications
| Length (Metric) | 4 m |
| Width (Metric) | 20 cm |
| For Use With (Application) | Western blots, dot-blot assays, and other protein or nucleic acid methods |
| Format | Roll |
| Material | 100% Pure Nitrocellulose |
| Pore Size | 0.2 μm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.