Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Amersham™ Protran™ Premium NC Nitrocellulose Membranes: Sheets

Catalog No. 45004038 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
10/Pk.
25/Pk.
5/Pk.
Length (Metric):
9 cm
10 cm
16 cm
20 cm
60 cm
Width (Metric):
8 cm
10 cm
14 cm
20 cm
30 cm
Pore Size:
0.2 μm
0.45 μm
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Length (Metric) Width (Metric) Pore Size
45-004-038 10/Pk. 20 cm 20 cm 0.45 μm
45-004-026 5/Pk. 60 cm 30 cm 0.45 μm
45-004-037 25/Pk. 20 cm 20 cm 0.45 μm
45-004-051 25/Pk. 16 cm 14 cm 0.45 μm
45-004-062 10/Pk. 10 cm 10 cm 0.45 μm
45-004-063 25/Pk. 10 cm 10 cm 0.2 μm
45-004-075 25/Pk. 9 cm 8 cm 0.45 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 45-004-038 Supplier Cytiva Supplier No. 10600048
Add to Cart
Add to Cart
This item is not returnable. View return policy

Use for high sensitivity, resolution, and low background for all labeling and detection systems, including chemiluminescent, fluorescenct, colorimetric and radioactive. Cytiva Amersham™ Protran™ Premium NC Nitrocellulose Membranes: Sheets offer excellent sensitivity, resolution, and low background.

Individual sheets.

 

  • Low background enables a dynamic range for complicated samples
  • Suitable for Western blots, dot-blot assays, and other protein or nucleic acid methods
  • 100% pure nitrocellulose
  • High affinity for proteins
  • Blocking ability

 

TRUSTED_SUSTAINABILITY

Specifications

Length (Metric) 20 cm
Width (Metric) 20 cm
For Use With (Application) Western blots, dot-blot assays, and other protein or nucleic acid methods
Quantity 10/Pk.
Format Sheet
Pore Size 0.45 μm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.